Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rac1,2,3 Mouse anti-Human, Clone: AT2G10, Novus Biologicals™

Mouse Monoclonal Antibody
$299.64 - $492.89
Specifications
| Antigen | Rac1,2,3 |
|---|---|
| Clone | AT2G10 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:1000, Flow Cytometry 2-5ug, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
Description
Rac1,2,3 Monoclonal specifically detects Rac1,2,3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Rac1,2,3 | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Recombinant human RAC2 (1-189aa) protein purified from E. coli | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| AT2G10 | |
| Western Blot 1:1000, Flow Cytometry 2-5ug, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 10% Glycerol | |
| IgG2b κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title