Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Antibody (CPTC-RAC1-1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB389210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389210 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389210 Supplier Novus Biologicals Supplier No. NBP308939UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rac1 Monoclonal antibody specifically detects Rac1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Rac1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-RAC1-1
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25
Host Species Mouse
Immunogen Recombinant human full-length Rac1protein (Uniprot: P63000)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5879
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.