Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10091266 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10091266 0.1 mL
NB017893 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10091266 Supplier Novus Biologicals Supplier No. NB10091266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Rac1 Polyclonal specifically detects Rac1 in Human, Mouse, Rabbit, Rat samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Rac1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:1000 - 1:2000, Immunohistochemistry-Frozen 1:1000 - 1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P63000
Gene Alias Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25
Gene Symbols RAC1
Host Species Rabbit
Immunogen This Rac1 antibody was developed against a synthetic peptide from amino acid region 100-150 as a part of human Rac1 conjugated to blue carrier protein.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5879
Test Specificity Detects both RAC1 (Gene ID: 5879, UniProt: P63000) and RAC2 (Gene ID: 5880, UniProt: P15153).
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse, Mouse, Rabbit, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.