Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rac1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$206.50 - $499.50
Specifications
Antigen | Rac1 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017893
![]() |
Novus Biologicals
NB100912660.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB10091266
![]() |
Novus Biologicals
NB10091266 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Rac1 Polyclonal specifically detects Rac1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Rac1 | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, mTOR Pathway | |
P63000 | |
5879 | |
IgG | |
Unpurified | |
Detects both RAC1 (Gene ID: 5879, UniProt: P63000) and RAC2 (Gene ID: 5880, UniProt: P15153). |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25 | |
RAC1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title