Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB325851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325851 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325851 Supplier Novus Biologicals Supplier No. NBP299370PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rac1 Polyclonal antibody specifically detects Rac1 in Human samples. It is validated for ELISA

Specifications

Antigen Rac1
Applications ELISA
Classification Polyclonal
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias Cell migration-inducing gene 5 protein, MGC111543, p21-Rac1migration-inducing gene 5, Ras-like protein TC25, ras-related C3 botulinum toxin substrate 1, ras-related C3 botulinum toxin substrate 1 (rho family, small GTP bindingprotein Rac1), rho family, small GTP binding protein Rac1, TC25, TC-25
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Rac1 (Accession#: NP_008839.2; Met1-Cys189)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5879
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.