Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rac1/CDC42, p Ser71 Antibody (SR1379), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.59 - $417.40
Specifications
| Antigen | Rac1/CDC42 |
|---|---|
| Clone | SR1379 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Rac1/CDC42 Monoclonal antibody specifically detects Rac1/CDC42 in Human samples. It is validated for Western BlotSpecifications
| Rac1/CDC42 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR1379 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A synthesized peptide derived from human Phospho-Rac1/CDC42 [Ser71] (Uniprot #: P60953, P63000) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title