Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1/CDC42, p Ser71 Antibody (SR1379), Novus Biologicals™
SDP

Catalog No. NB258692 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258692 100 μg
NB258454 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258692 Supplier Novus Biologicals Supplier No. NBP322649100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rac1/CDC42 Monoclonal antibody specifically detects Rac1/CDC42 in Human samples. It is validated for Western Blot

Specifications

Antigen Rac1/CDC42
Applications Western Blot
Classification Monoclonal
Clone SR1379
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Host Species Rabbit
Immunogen A synthesized peptide derived from human Phospho-Rac1/CDC42 [Ser71] (Uniprot #: P60953, P63000)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.