Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAC3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB331554 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331554 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331554 Supplier Novus Biologicals Supplier No. NBP297482MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAC3 Polyclonal antibody specifically detects RAC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen RAC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias p21-Rac3, ras-related C3 botulinum toxin substrate 3, ras-related C3 botulinum toxin substrate 3 (rho family, small GTP bindingprotein Rac3), rho family, small GTP binding protein Rac3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RAC3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5881
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.