Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB046989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046989 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046989 Supplier Novus Biologicals Supplier No. NBP297717F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rad1 Polyclonal specifically detects Rad1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rad1
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias cell cycle checkpoint protein Hrad1, checkpoint control protein HRAD1, DNA repair exonuclease rad1 homolog, DNA repair exonuclease REC1, EC 3.1.11.2, exonuclease homolog RAD1, HRAD1, RAD1 (S. pombe) homolog, RAD1 homolog (S. pombe), Rad1-like DNA damage checkpoint protein, REC1cell cycle checkpoint protein RAD1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Rad1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5810
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.