Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad17 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000929 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000929 Supplier Novus Biologicals Supplier No. NB100173SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Rad17 Polyclonal specifically detects Rad17 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen Rad17
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 -1:1000, Flow Cytometry
Formulation PBS with 0.01% Sodium Azide
Gene Alias CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog
Gene Symbols RAD17
Host Species Rabbit
Immunogen Synthetic peptide made to a N-terminal region of human RAD17.
Molecular Weight of Antigen 66 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5884
Test Specificity Reacts with residues 2-15 [NQVTDWVDPSFDDF] of the human RAD17 protein.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.