Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad17, p Ser645 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292267 Supplier Novus Biologicals Supplier No. NB100273APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rad17 Polyclonal specifically detects Rad17 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen Rad17
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias CCYC, cell cycle checkpoint protein (RAD17), cell cycle checkpoint protein RAD17, FLJ41520, HRAD17, R24L, RAD1 (S. pombe) homolog, RAD1 homolog, RAD17 homolog (S. pombe), Rad17-like protein, RAD17SP, Rad24, RF-C activator 1 homolog, RF-C/activator 1 homolog
Host Species Rabbit
Immunogen A synthetic peptide, which represented a portion of human Rad17 (LocusLink ID 5884) around serine 645.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5884
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.