Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD18 Antibody (79B1048.1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB339627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339627 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339627 Supplier Novus Biologicals Supplier No. NB10056523AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAD18 Monoclonal antibody specifically detects RAD18 in Human samples. It is validated for Western Blot

Specifications

Antigen RAD18
Applications Western Blot
Classification Monoclonal
Clone 79B1048.1
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, hHR18, hRAD18, postreplication repair protein hRAD18p, Postreplication repair protein RAD18, RAD18 homolog (S. cerevisiae), RAD18, S. cerevisiae, homolog, RING finger protein 73, RNF73E3 ubiquitin-protein ligase RAD18
Host Species Mouse
Immunogen The antibody was raised against a synthetic peptide corresponding to the amino acids 402-414 of human Rad18.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 56852
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.