Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad21 Antibody (CM110-2C10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB10386A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10386A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10386A350 Supplier Novus Biologicals Supplier No. NB100386AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad21 Monoclonal specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Rad21
Applications Western Blot
Classification Monoclonal
Clone CM110-2C10
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596
Gene Symbols RAD21
Host Species Mouse
Immunogen An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5885
Test Specificity NB100-386 is specific for both the nuclear hRad21 protein and the cleaved cytoplasmic hRad21 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.