Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad21 Antibody (CM110-2C10), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB339717 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339717 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339717 Supplier Novus Biologicals Supplier No. NB100386MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad21 Monoclonal specifically detects Rad21 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Rad21
Applications Western Blot
Classification Monoclonal
Clone CM110-2C10
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western
Formulation 50mM Sodium Borate
Gene Alias double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596
Host Species Mouse
Immunogen An N-terminal fusion protein of the C-terminal fragment of Scc1 subunit human cohesin (amino acids 544-631 of hRad21).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5885
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.