Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad21 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220053 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220053 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220053 Supplier Novus Biologicals Supplier No. NBP220053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rad21 Polyclonal specifically detects Rad21 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Rad21
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:100-1:500
Formulation PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300
Gene Accession No. O60216
Gene Alias double-strand-break repair protein rad21 homolog, hHR21FLJ25655, HR21, KIAA0078RAD21 (S. pombe) homolog, MCD1, Nuclear matrix protein 1, NXP-1, NXP1HRAD21, protein involved in DNA double-strand break repair, RAD21 homolog (S. pombe), SCC1 homolog, SCC1FLJ40596
Gene Symbols RAD21
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAD21. The exact sequence is proprietary.
Molecular Weight of Antigen 72 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5885
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.