Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad21 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7139968
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-6856 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-6856 Supplier Bethyl Laboratories, Inc Supplier No. A300080AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

The cohesin complex plays an important role in tethering sister chromatids during the prophase through anaphase stages of mitosis, making certain that genomic information is replicated accurately. As the cellular division process continues, separase destroys the cohesin complex by means of cleavaging its Scc1 subunit, allowing the chromatids to separate and divide with the cell. During apoptosis the C-terminal portion of hRad21 is cleaved and translocated to the cytoplasm. This cleavage and translocation may act as a nuclear signal that promotes and accelerates subsequent events of apoptosis.

Specifications

Antigen Rad21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RAD21
Gene Accession No. O60216, Q61550
Gene Alias double-strand-break repair protein rad21 homolog, hHR21, HR21, KIAA0078RAD21, MCD1, Nuclear matrix protein 1, NXP-1, HRAD21, protein involved in DNA double-strand break repair, SCC1 homolog
Gene Symbols Rad21
Host Species Rabbit
Immunogen Between 575 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19357, 5885
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.