Learn More
Description
Specifically detects Rad3 in Yeast samples, and it is validated for Western Blotting
Specifications
Specifications
| Antigen | Rad3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of Saccharomyces cerevisiae S288c RAD3. The exact sequence is proprietary. |
| Molecular Weight of Antigen | 90 kDa |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
