Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad50 Antibody, Novus Biologicals™
SDP

Catalog No. NB100154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100154 0.1 mL
NB100154SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100154 Supplier Novus Biologicals Supplier No. NB100154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rad50 Polyclonal specifically detects Rad50 in Hamster, Human, Mouse samples. It is validated for ICC/IF (Negative), Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vivo Assay, Simple Western, Western Blot.

Specifications

Antigen Rad50
Applications ICC/IF (Negative), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vivo Assay, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, Simple Western 1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, ICC/IF (Negative)
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2
Gene Symbols RAD50
Host Species Rabbit
Immunogen C-terminal mouse RAD50 (604 amino acids). [UniProt# P70388]
Molecular Weight of Antigen 153 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 10111
Target Species Hamster, Mouse, Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.