Learn More
Description
Specifications
Specifications
| Antigen | Rad50 |
| Applications | ICC/IF (Negative), Immunocytochemistry, Immunofluorescence, Immunoprecipitation, In vivo Assay, Simple Western, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, Simple Western 1:2000, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 1:10-1:500, ICC/IF (Negative) |
| Formulation | Whole antisera with 0.02% Sodium Azide |
| Gene Alias | DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 |
| Gene Symbols | RAD50 |
| Host Species | Rabbit |
| Immunogen | C-terminal mouse RAD50 (604 amino acids). [UniProt# P70388] |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
