Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$181.72 - $492.89
Specifications
| Antigen | Rad50 |
|---|---|
| Dilution | Western Blot 1:1000-1:10000, Simple Western 1:1000, Immunoprecipitation 1-5ul |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1001487SS
![]() |
Novus Biologicals
NB1001487SS |
0.025 mL |
Each for $181.72
|
|
|||||
NB1001487
![]() |
Novus Biologicals
NB1001487 |
0.1 mL |
Each for $492.89
|
|
|||||
Description
Rad50 Polyclonal specifically detects Rad50 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
| Rad50 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining | |
| Whole antisera with No Preservative | |
| DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2 | |
| RAD50 | |
| IgG | |
| Unpurified | |
| 153 kDa |
| Western Blot 1:1000-1:10000, Simple Western 1:1000, Immunoprecipitation 1-5ul | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q92878 | |
| 10111 | |
| Human RAD50 zinc hook region (amino acids 628-787) expressed in E. coli. [UniProt# Q92878] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title