Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad50 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001487SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001487SS 0.025 mL
NB1001487 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001487SS Supplier Novus Biologicals Supplier No. NB1001487SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Rad50 Polyclonal specifically detects Rad50 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.

Specifications

Antigen Rad50
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Simple Western 1:1000, Immunoprecipitation 1-5ul
Formulation Whole antisera with No Preservative
Gene Accession No. Q92878
Gene Alias DNA repair protein RAD50, EC 3.6, EC 3.6.1.15, EC 3.6.3, EC 3.6.3.27, hRAD50, NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog (S. cerevisiae), RAD502, RAD50-2
Gene Symbols RAD50
Host Species Rabbit
Immunogen Human RAD50 zinc hook region (amino acids 628-787) expressed in E. coli. [UniProt# Q92878]
Molecular Weight of Antigen 153 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 10111
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.