Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51 Antibody (14B4), Novus Biologicals™
SDP

Catalog No. NB100148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100148 Supplier Novus Biologicals Supplier No. NB100148
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 52 publications

Rad51 Monoclonal specifically detects Rad51 in Caenorhabditis elegans, Chicken, Human, Mouse, Rat samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, Proximity Ligation Assay, Western Blot.

Specifications

Antigen Rad51
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vitro Assay, KnockDown, Proximity Ligation Assay, Western Blot
Classification Monoclonal
Clone 14B4
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:3000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100 - 1:1000, Immunohistochemistry-Frozen, In vitro assay, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP), Knockdown Validated
Gene Accession No. Q06609
Gene Alias BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
Gene Symbols RAD51
Host Species Mouse
Immunogen Full length (amino acids 1-338) Rad51 expressed in E. coli.
Molecular Weight of Antigen 37 kDa
Purification Method Antigen Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 5888
Test Specificity Reactivity with C.elegans reported in scientific literature (PMID: 23942865). Mouse reactivity reported in scientific literature (PMID: 25218637). RAD51 is highly conserved in eukaryotes and this antibody may have wide range of species reactivity.
Target Species C. elegans, Chicken, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.