Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51 Antibody (523713), HRP, R&D Systems™
SDP

Catalog No. NB003390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB003390 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB003390 Supplier Novus Biologicals Supplier No. MAB99481H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad51 Monoclonal specifically detects Rad51 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Rad51
Applications Immunohistochemistry
Classification Monoclonal
Clone 523713
Conjugate HRP
Dilution Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RAD51L3, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
Gene Symbols RAD51
Host Species Mouse
Immunogen E. coli-derived partial recombinant human Rad51 protein (amino acids 174-339)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies, Phospho Specific
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.