Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51C Antibody (2H11/6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB10177A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10177A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10177A532 Supplier Novus Biologicals Supplier No. NB100177AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Rad51C
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H11/6
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3
Gene Symbols RAD51C
Host Species Mouse
Immunogen His-tagged human Rad51C, over-expressed in E. coli. [UniProtNo. O43502]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5889
Test Specificity Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis.
Target Species Human, Mouse, Primate, Yeast
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.