Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51C Antibody (2H11/6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB100177IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100177IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100177IR Supplier Novus Biologicals Supplier No. NB100177IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad51C Monoclonal specifically detects Rad51C in Human, Mouse, Primate, Yeast samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Rad51C
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H11/6
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. O43502
Gene Alias BROVCA3, DNA repair protein RAD51 homolog 3, FANCO, MGC104277, R51H3, RAD51 homolog C, RAD51 homolog C (S. cerevisiae), RAD51L2RAD51 (S. cerevisiae) homolog C, RAD51-like protein 2, yeast RAD51 homolog 3
Gene Symbols RAD51C
Host Species Mouse
Immunogen His-tagged human Rad51C, over-expressed in E. coli. [UniProt# O43502]
Molecular Weight of Antigen 40 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 5889
Test Specificity Does not cross-react with Rad51B, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis.
Target Species Human, Mouse, Primate, Yeast
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.