Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51D Antibody (5B3/6), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 3 publications

Supplier:  Novus Biologicals NB100178

 View more versions of this product

Catalog No. NB100178


Only null left
Add to Cart

Description

Description

Rad51D Monoclonal specifically detects Rad51D in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Rad51D
Monoclonal
Unconjugated
Ascites with 0.1% Sodium Azide
DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD
Mouse
40 kDa
0.1 mL
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
5892
Human, Mouse (Negative)
Ascites
Western Blot, Immunocytochemistry, Immunofluorescence
5B3/6
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500
O75771
RAD51D
His-tagged human Rad51D, overexpressed in E. coli. [UniProt# O75771]
Unpurified
RUO
Primary
Human. This antibody does not work on mouse.
Store at 4C. Do not freeze.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only