Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51D Antibody (5B3/6), Novus Biologicals™
SDP

Catalog No. p-4998633 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100178SS 0.025 mL
NB100178 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100178SS Supplier Novus Biologicals Supplier No. NB100178SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Rad51D Monoclonal specifically detects Rad51D in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rad51D
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5B3/6
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. O75771
Gene Alias DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD
Gene Symbols RAD51D
Host Species Mouse
Immunogen His-tagged human Rad51D, overexpressed in E. coli. [UniProt# O75771]
Molecular Weight of Antigen 40 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 5892
Test Specificity Human. This antibody does not work on mouse.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C. Do not freeze.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.