Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51D Antibody (Rad51D 5A8/4), PE, Novus Biologicals™
SDP

Catalog No. NB309663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309663 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309663 Supplier Novus Biologicals Supplier No. NBP250264PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad51D Monoclonal antibody specifically detects Rad51D in Human, Hamster samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Rad51D
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone Rad51D 5A8/4
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias DNA repair protein RAD51 homolog 4, HsTRAD, R51H3Trad, RAD51 homolog D, RAD51DRAD51 (S. cerevisiae)-like 3, RAD51-like 3 (S. cerevisiae), RAD51-like protein 3, recombination repair protein, TRAD
Host Species Mouse
Immunogen Human recombinant denatured protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 5892
Target Species Human, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.