Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD51D Monoclonal Antibody (5B3/6)
GREENER_CHOICE

Catalog No. AMPA6542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMPA6542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMPA6542 Supplier Invitrogen™ Supplier No. MA116542
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: antigen standard for RAD51L3 (transient overexpression lysate). This antibody does not work on mouse.

Five human RAD 51 homologues have been identified: XRCC2, XRCC3, RAD51B, RAD51C, and RAD51D. Each of these homologues interacts with one or more of the others, with all of the proteins involved in one complex or multiple smaller complexes. RAD51D interacts with XRCC2 and RAD51C. RAD51D's expression pattern and sequence similarity to other RAD51 family members most likely make it part of a complex of proteins required for DNA repair and meiotic recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD51D
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5B3/6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene RAD51D
Gene Accession No. O55230, O75771
Gene Alias BROVCA4; DNA repair protein RAD51 homolog 4; DNA repair protein RAD51-like 4; HsTRAD; R51H3; RAD51 homolog D; RAD51 paralog D; Rad51d; RAD51L3; RAD51-like 3; RAD51-like protein 3; recombination repair protein; TRAD
Gene Symbols RAD51D
Host Species Mouse
Immunogen His-tagged human Rad51D, overexpressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19364, 5892
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.