Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51L1 Antibody (1 H3/13) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100176 0.1 mL
NB100176SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100176 Supplier Novus Biologicals Supplier No. NB100176
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Rad51L1 Monoclonal specifically detects Rad51L1 in Human, Mouse (Negative), Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen Rad51L1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1 H3/13
Concentration 0.7 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature
Formulation PBS with 0.1% Sodium Azide
Gene Accession No. O15315
Gene Alias DNA repair protein RAD51 homolog 2, MGC34245, R51H2REC2hREC2, RAD51 (S. cerevisiae)-like 1, RAD51 homolog B, RAD51B, RAD51-like 1 (S. cerevisiae), RAD51-like protein 1, RecA-like protein, recombination repair protein
Gene Symbols RAD51B
Host Species Mouse
Immunogen His-tagged human Rad51L1, overexpressed in E. coli. [UniProt# O15315]
Molecular Weight of Antigen 42 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 5890
Test Specificity Does not cross-react with Rad51C, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis.
Target Species Human, Primate, Mouse (Negative), Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.