Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51L1 Antibody (1 H3/13) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | Rad51L1 |
---|---|
Clone | 1 H3/13 |
Concentration | 0.7 mg/ml |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100176SS
![]() |
Novus Biologicals
NB100176SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB100176
![]() |
Novus Biologicals
NB100176 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Rad51L1 Monoclonal specifically detects Rad51L1 in Human, Primate, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Rad51L1 | |
0.7 mg/ml | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination | |
PBS with 0.1% Sodium Azide | |
DNA repair protein RAD51 homolog 2, MGC34245, R51H2REC2hREC2, RAD51 (S. cerevisiae)-like 1, RAD51 homolog B, RAD51B, RAD51-like 1 (S. cerevisiae), RAD51-like protein 1, RecA-like protein, recombination repair protein | |
RAD51B | |
IgG2a | |
Protein A or G purified | |
Does not cross-react with Rad51C, Rad51D, Rad51, XRCC2, or XRCC3 in Western analysis. |
1 H3/13 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence reported in scientific literature | |
Monoclonal | |
Ascites | |
RUO | |
Human, Primate, Mouse (Negative) | |
O15315 | |
5890 | |
His-tagged human Rad51L1, overexpressed in E. coli. [UniProt# O15315] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title