Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD54L Antibody (38.4), Novus Biologicals™
SDP

Catalog No. NBP141218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141218 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141218 Supplier Novus Biologicals Supplier No. NBP141218
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAD54L Monoclonal specifically detects RAD54L in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen RAD54L
Applications Western Blot, ELISA
Classification Monoclonal
Clone 38.4
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Gene Accession No. Q92698
Gene Alias EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae)
Gene Symbols RAD54L
Host Species Mouse
Immunogen Synthetic peptide - Combination of N & C peptide - Corresponding to a range between amino acids 1-300 and 450-741.
Purification Method Immunogen affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8438
Test Specificity This antibody will recognize human Rad 54.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.