Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD54L Antibody (Rad54 4E3/1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB11013364N Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11013364N 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11013364N Supplier Novus Biologicals Supplier No. NB11013364AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAD54L Monoclonal specifically detects RAD54L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RAD54L
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone Rad54 4E3/1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.6.1, hHR54EC 3.6.4.-, HR54, hRAD54HRAD54, RAD54 (S.cerevisiae)-like, RAD54 homolog, RAD54ADNA repair and recombination protein RAD54-like, RAD54-like (S. cerevisiae)
Gene Symbols RAD54L
Host Species Mouse
Immunogen Full length human RAD54L protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Homologous Recombination
Primary or Secondary Primary
Gene ID (Entrez) 8438
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.