Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad9 Antibody (93A535), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1201360A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1201360A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1201360A70 Supplier Novus Biologicals Supplier No. NB12013600AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Rad9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 93A535
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe)
Gene Symbols RAD9A
Host Species Mouse
Immunogen The antibody was raised against fusion protein of human full length Rad9.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Checkpoint signaling, Chromatin Research, DNA Repair, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5883.0
Test Specificity In recombinant Rad9, a ∼50 kDa band is observed.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.