Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD9B Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB293618 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293618 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293618 Supplier Novus Biologicals Supplier No. NBP326072AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAD9B Polyclonal antibody specifically detects RAD9B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen RAD9B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias cell cycle checkpoint control protein RAD9B, cell cycle checkpoint control protein RAD9B homolog, DNA repair exonuclease rad9 homolog B, FLJ40346, hRAD9B, MGC75426, RAD9 homolog B (S. pombe)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RAD9B fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 144715
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.