Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAD9B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113016
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5113016 100 μg
1 options

Catalog No. PIPA5113016

Supplier: Invitrogen™ PA5113016

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAD9B Polyclonal Antibody for Western Blot, ELISA

RAD9B gene ontology (GO) annotations include DNA repair; DNA replication; cellular response to ionizing radiation; intra-S DNA damage checkpoint; regulation of signal transduction by p53 class mediator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAD9B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RAD9B
Gene Accession No. Q6WBX7, Q6WBX8
Gene Alias A630082N15Rik; BC021784; Cell cycle checkpoint control protein RAD9B; cell cycle checkpoint control protein RAD9B homolog; DNA repair exonuclease rad9 homolog B; hRAD9B; mRAD9B; RAD9 checkpoint clamp component B; RAD9 homolog B; RAD9B
Gene Symbols RAD9B
Host Species Rabbit
Immunogen Recombinant Human Cell cycle checkpoint control protein RAD9B protein (274-408AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144715, 231724
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.