Learn More
United Scientific Supplies Radiation Can Kit
SDPSCI_ED
Catalog No. p-4661123 Shop All United Scientific Supplies, Inc Products
Click to view available options
Product Type:
Lamp Assembly
Radiation Can Kit
This item is not returnable. View return policy
Catalog No. S07396
Supplier: United Scientific Supplies LWB001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Lamp assembly with reflector and stand (bulb not included), thermometer, one smooth metal can, one black metal can, thermometer.
Demonstrate how two identically sized cans absorb and radiate energy differently.
  • Includes two cans: one smooth metal can w/screw cap and other black metal can
  • Thermal radiation emitted by two cans cools or heats water at different rates
Order Info
Lamp assembly with stand is also sold separately.
Specifications
For Use With (Equipment) Accessory for radiation can kit
Includes Lamp assembly with reflector and stand (bulb not included), thermometer, one smooth Metal can, one black Metal can, thermometer
Product Type Lamp Assembly
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.