Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ radiello™ Passive Diffusive Air Samplers Starter Kit
Shop All Restek Corporation Products

Click to view available options
Type:
Complete sampler for 1,3-butadiene & isoprene, thermal desorption, yellow diffusive body, cartridge + blank, support plate, adaptor
Complete sampler for BTEX/VOCs, CS2 desorption, white diffusive body, cartridge + blank, support plate, adaptor
Complete sampler for BTEX/VOCs, thermal desorption, yellow diffusive body, cartridge + blank, support plate, adaptor
Complete sampler for H2S, white diffusive body, cartridge + blank, support plate, adaptor
Description
- Versatile radiello™ passive diffusive air samplers—now available from Restek™ —feature a unique 3D radial design that provides several performance advancements compared to standard 2D flat axial air samplers
- These samplers are a reliable, cost-effective alternative that offers both high sensitivity for low-level detection in short sampling periods (down to 15 min), as well as higher capacity so accurate results can be obtained over longer (30 day) sampling times
- Because the radial design results in a higher rate of uptake and larger capacity, radiello™ air samplers are a versatile solution that allows labs to sample very efficiently for short durations or sample for longer periods without bias from analyte breakthrough or back diffusion
- These air samplers are available for a wide range of chemical classes, so the choice of sampling material can be tailored to specific applications
- Ideal for labs supporting environmental monitoring, industrial hygiene, and any other ambient (indoor/outdoor) air testing, radiello™ air samplers from Restek are recommended when a reliable source of versatile, high-performing, easy-to-use air samplers is required
- Solvent and thermal desorption options available
- Features water repellant diffusive body and reusable holders
- Easy to use—no pump required
Specifications
Specifications
Material | Mixed |
For Use With (Application) | Personal Air Sampling |
Quantity | 1 |
Type | Complete sampler for BTEX/VOCs, CS2 desorption, white diffusive body, cartridge + blank, support plate, adaptor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction