Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rae-1 Antibody (199215), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB175367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175367 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175367 Supplier Novus Biologicals Supplier No. FAB1758Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Rae-1 Monoclonal specifically detects Rae-1 in Mouse samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry.

Specifications

Antigen Rae-1
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry
Classification Monoclonal
Clone 199215
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with Rae-1 beta and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8480
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.