Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAE1 delta Monoclonal Antibody (RD-41), eBioscience™

Catalog No. p-7177422
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-131-30 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-131-30 Supplier LIFE TECHNOLOGIES Supplier No. 14575681
Only null left
Add to Cart
Add to Cart

RAE1 delta Monoclonal Antibody (RD-41), eBioscience™

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAE1 delta
Applications ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RD-41
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Raet1d
Gene Accession No. Q9JI58
Gene Alias RAE-1delta; RAE-1-delta; Raet1d; retinoic acid early transcript 1D; retinoic acid early transcript delta; retinoic acid early-inducible protein 1-delta
Gene Symbols Raet1d
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56554
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.