Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Raf-1, p Ser621 Antibody (SR2183), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Raf-1 |
---|---|
Clone | SR2183 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Raf-1 Monoclonal antibody specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Raf-1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
A synthesized peptide derived from human phospho-Raf-1 [Ser621] (Uniprot #: P04049) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR2183 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5894 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title