Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raf-1 Rabbit anti-Human, Clone: SD087-05, Novus Biologicals™
SDP

Catalog No. NBP267808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267808 Supplier Novus Biologicals Supplier No. NBP267808

Rabbit Monoclonal Antibody

Raf-1 Monoclonal specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Raf-1
Applications Western Blot
Classification Monoclonal
Clone SD087-05
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1
Gene Symbols RAF1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser43 of Human Raf-1 aa 20-69/ 648. (SwissProt: P04049 Human; SwissProt: Q99N57 Mouse; SwissProt: P11345 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5894
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.