Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAG1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP174190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174190 100 μL
NBP17419020 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP174190 Supplier Novus Biologicals Supplier No. NBP174190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

RAG1 Polyclonal specifically detects RAG1 in Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.

Specifications

Antigen RAG1
Applications Western Blot, ChIP Assay
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P15919
Gene Alias MGC43321, RAG-1, recombination activating gene 1, RING finger protein 74, RNF74recombination activating protein 1, V(D)J recombination-activating protein 1
Gene Symbols RAG1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the C terminal of Rag1. Immunizing peptide sequence IIERDGSIGAWASEGNESGNKLFRRFRKMNARQSKCYEMEDVLKHHWLYT.
Molecular Weight of Antigen 119 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5896
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Mouse, Rat, Pig, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.