Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAG1AP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001257 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001257 Supplier Novus Biologicals Supplier No. NB1001257
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RAG1AP1 Polyclonal specifically detects RAG1AP1 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen RAG1AP1
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_061333.
Gene Alias HsSWEET1RAG1-activating protein 1, probable sugar transporter RAG1AP1, RAG1AP1SWEET1, recombination activating gene 1 activating protein 1, RZPDo834D038D, SCPRP11-540D14.5, slv, solute carrier family 50 (sugar transporter), member 1, Solute carrier family 50 member 1, Stromal cell protein
Gene Symbols SLC50A1
Host Species Goat
Immunogen Peptide with sequence C-EQDRNYWLLQT corresponding to C-Terminus according to NP_061333.2, NP_001116309.1, NP_001116311.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55974
Test Specificity This antibody is expected to recognize the reported isoforms.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.