Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAGEF2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106550 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106550 Supplier Novus Biologicals Supplier No. NBP106550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAGEF2 Polyclonal specifically detects RAGEF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen RAGEF2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_057424.2
Gene Alias DKFZp667N084, DKFZp686I15116, KIA001LB, PDZ domain-containing guanine nucleotide exchange factor 2, PDZ domain-containing guanine nucleotide exchange factor I, PDZ-GEF2RA-GEF-2PDZGEF2PDZ domain containing guanine nucleotide exchange factor (GEF) 2, RAGEF2, Rap guanine nucleotide exchange factor (GEF) 6, rap guanine nucleotide exchange factor 6
Gene Symbols RAPGEF6
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human PDZ domain containing guanine nucleotide exchange factor (GEF) 2 using the numbering given in entry NP_057424.2 (GeneID 51735).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51735
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.