Learn More
Description
Specifications
Specifications
| Antigen | RAI1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000-1:10000, Immunohistochemistry 1:500-1:2000, Immunoprecipitation 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Formulation | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gene Accession No. | NP_109590.3 |
| Gene Alias | KIAA1820DKFZP434A139, MGC12824, retinoic acid induced 1, retinoic acid-induced protein 1, SMCR, Smith-Magenis syndrome chromosome region, SMSDKFZp434A139 |
| Gene Symbols | RAI1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
