Learn More
Description
Specifications
Specifications
| Antigen | RAI16 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | family with sequence similarity 160, member B2, FLJ11125, FLJ21801, MGC138352, RAI16, retinoic acid induced 16, Retinoic acid-induced protein 16 |
| Gene Symbols | FHIP2B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human RAI16 fragment. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
