Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAIDD/CRADD Antibody (14G8), HRP, Novus Biologicals™
SDP

Catalog No. NBP242688H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242688H 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242688H Supplier Novus Biologicals Supplier No. NBP242688H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAIDD/CRADD Monoclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen RAIDD/CRADD
Applications Western Blot, ELISA
Classification Monoclonal
Clone 14G8
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Gene Alias CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain
Gene Symbols CRADD
Host Species Mouse
Immunogen Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8738
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.