Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAIDD/CRADD Antibody (14G8), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309909 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309909 Supplier Novus Biologicals Supplier No. NBP242688MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAIDD/CRADD Monoclonal antibody specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen RAIDD/CRADD
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 14G8
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain
Host Species Mouse
Immunogen Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8738
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.