Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RALY Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB102867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102867 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102867 Supplier Novus Biologicals Supplier No. NBP297634DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RALY Polyclonal specifically detects RALY in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RALY
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias Autoantigen p542, Heterogeneous nuclear ribonucleoprotein C-like 2, hnRNP associated with lethal yellow protein homolog, hnRNP core protein C-like 2, HNRPCL2MGC117312, P542hnRNP-associated with lethal yellow), RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow), RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog(mouse)), RNA-binding protein (autoantigenic), RNA-binding protein Raly
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RALY fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 22913
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.